Learn More
Thermo Scientific 24-Place Fixed-Angle Rotors with Covers
$2497.00
Catalog Number | Mfr. No. | Angle | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Angle | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
75-007-593
|
Thermo Scientific
75007593 |
45° | 21,890 x g | 15,000rpm |
Each for $2,497.00
|
|
75-007-599
|
Thermo Scientific
75007599 |
40° | 16,060 x g | 13,000rpm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Specifications
24 x 1.5/2mL | |
Microliter Rotor |