Learn More
Deltatrak IncSupplier Diversity Partner 24006 ISFET POCKET PH METER, Packaged individually DFS Item
Manufacturer:  Deltatrak IncSupplier Diversity Partner 24006
Model 24006 is a low-cost non-glass Pocket ISFET (Ion-Sensitive Field Effect Transistor) pH Meter featuring a virtually unbreakable silicon chip sensor that gives stable readings in seconds. The pH meter is rated IP67 waterproof and is ideal in applications involving food and beverage, medical, pharmaceutical, agriculture, and environmental studies. The ISFET pH meter is capable of reading a single drop sample and stores dry requiring no storage solution. Sensor Silicon chip ISFET* pH sensor with on-chip temperature sensor Replaceable reference electrode Calibration Capable of one or two point calibration Automatic Buffer Recognition Standard buffers of pH 4.0, 7.0, and 9.2 Display LCD with pH and Temperature pH Range: 2.0 to 12.0 pH Resolution: 0.01 pH Accuracy: 0.1pH Temperature Operating Temperature Range: 41F to 104F (5C to 40C) Measurement Range: 32F to 122 F (0C to 50 C) Resolution: 0.1C or 0.1F Accuracy: 2.0F / 1.0C
Catalog No. NC1478483