Learn More
25-OH Vitamin D3 Rabbit anti-Not Applicable, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Manufacturer:  Invitrogen MA527880
Catalog No. PIMA527880
Add to cart
Description
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using Recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
25-OH Vitamin D3 (calcifediol) is a prohormone produced by the liver by hydroxylation of vitamin D3 and is used to measure vitamin D status and detect itamin D deficiency.
Specifications
Specifications
25-OH Vitamin D3
Recombinant Monoclonal
Unconjugated
Liquid
25-OH Vitamin D3 BSA conjugate
Protein A
RUO
Antibody
Monoclonal
ELISA
RM3
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Rabbit
IgG
100 μl
-20°C
Primary
Not Applicable