Learn More
Leap PAL PartsSupplier Diversity Partner L-MARK Brand 250l Gold Syringe, FN, GT, 22g, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner LMK.2620821C
L-MARK Brand Syringe, 250uL, FN, Chemically Inert Coated Needle, GT, PTFE, 22g, Pt. 3, (Barrel 6.6mm); Syringe Adapter: PAL.SA250A Made in Germany.
Catalog No. 50-635-358