Learn More
Leap PAL Parts L-MARK Brand 250l, Collect PAL RN, NA, PTFE, Pt. Not, Sidehole #10-32
Supplier: Leap PAL Parts LMK.2620828
L-MARK Brand 250l, L-MARK Brand Syringe Collect PAL RN, GT, 0.15mm i.d. Plunger, For Low Flow (up to 250-300 l/min) applications NA Gauge, NDL Length , Pt. Style Not Included, Plunger Tip PTFE, Single Made in Germany.