Learn More
Cole-Parmer Pipette Bulb, natural rubber and silicone, 25 mL capacity  DFS Item
Manufacturer:  Cole-Parmer 2482060
These natural rubber bulbs are sturdy, re-usable and easy-to-use can be used with Pasteur, serological, volumetric and other standard pipettes. The bulbs have a silicone seal to grip the glass firmly and protect it.
Catalog No. NC9909042