Learn More
MilliporeSigma™ Accessories for Glass Microanalysis Filter Holders
Accessories for filter holders for sample preparation.
Supplier:  MilliporeSigma™ XX1012532
 View more versions of this product
Catalog No. XX1012532
Add to Cart
Description
Description
Accessories for filter holders for sample preparation.
Millipore Glass Vacuum Filtration Accessory ProductThese accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration. This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits.
Specifications
Specifications
Not Autoclavable
25 mm glass microanalysis vacuum filter
Non-sterile
25 mm
Base, stopper and screen
Borosilicate Glass
Base, Stopper and Screen
Product Suggestions
Product Suggestions