Learn More
Leap PAL PartsSupplier Diversity Partner L-MARK Brand Syringe, 25l, FN, GT, PTFE, 22gs, Pt. 3, Barrell Outer Diameter 7.8mm  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner LMK.2620530
L-MARK Brand Chromatography Syringes, Syringe, 25uL, FN, GT, PTFE, 22gs, Pt. 3, Barrell Outer Diameter 7.8mm Made in Germany.
Catalog No. 50-635-299