Learn More
Tele-Lite™ Honda™ High-Pressure Pumps
Easy to start, prime and maintain
$878.00 - $912.00
Specifications
Horsepower 5.5 hp
View More Specs
Products 2
Catalog Number Mfr. No. Horsepower Price Quantity & Availability  
Catalog Number Mfr. No. Horsepower Price Quantity & Availability  
19-220-440
View Documents
Tele-Lite™
WH15XK2A
4 hp
Each for $878.00
Add to cart
19-220-441
View Documents
Tele-Lite™
WH20XK2AC1
5.5 hp
Each for $912.00
Add to cart
Description
Description
High-pressure centrifugal self-priming water pumps offer high pressure with lower flow and are best suited for applications where higher pressure is required, such as when using sprinklers or a nozzle.
  • Transistorized pointless ignition systems are virtually maintenance free and easy to start under all conditions
  • Lightweight aluminum pump housing reduces overall pump weight
  • Inlet flapper valve allows pump case to be filled without water flowing out of the suction opening to reduce priming time and prevent siphoning after the engine is shut down
  • Corrosion-resistant pump case drain and fill plugs don't swell and are easily removed with fingers
  • Heavy-duty, rigid-mounted cast iron impeller and volute resist wear
  • Durable and easy-to-service ceramic-carbon mechanical water seal
  • Full tubular frame protects pump from damage and provides convenient lifting points
  • Powerful four-stroke engine operates quietly and is virtually smoke-free
  • Runs on regular gasoline, eliminating the hassle of mixing oil and gas
Compliance
Meet U.S. EPA and California Air Resources Board emission levels.
Specifications
Specifications
5.5 hp