Learn More
2B4/CD244/SLAMF4 Mouse anti-Human, Janelia Fluor 549, Clone: 2B4.69, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10065889JF549
Catalog No. NB065889549
Add to cart
Description
Description
2B4/CD244/SLAMF4 Monoclonal antibody specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
2B4/CD244/SLAMF4
2B4.69
Flow Cytometry
Purified
CD244
CD244 Fusion protein.
Protein G purified
Myeloid derived Suppressor Cell
Primary
51744
Human
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
NB100-65889 is specific for the CD244 cell surface antigen, also known as natural killer receptor 2B4 (NKG2B4). The glycoprotein CD244 is a member of the SLAM family of receptors which is expressed on natural killer (NK) cells, gamma delta T cells, CD14+ monocytes, and a subset of CD8+ T cells. CD244 was originally identified as an activating receptor on NK cells, although recent studies suggest that the receptor may have a wide range of activities including NK cell activation, costimulation, or even inhibition, depending on the cell type and stage of differentiation. The ligand of 2B4 is CD48, a GPI-anchored membrane protein also belonging to the SLAM family.