Learn More
Cole-Parmer Partlow 741200000021 10" Chart Recorder; 2 Channel/2 Relay  DFS Item
Manufacturer:  Cole-Parmer 4900190
Proven recording reliability that became the industry standard! Capable of measuring, displaying, recording and controlling up to two process variables from a variety of inputs. All user entered data can be protected against unauthorized changes. Straightforward programming allows you to configure your recorder with a logical step-by-step process using a simple keypad.
Catalog No. 50-628-409