Learn More
Cole-Parmer Monarch DC1250-U11 2ch Recorder With Universal Ac Rechargeable Battery Ups & USB  DFS Item
Manufacturer:  Cole-Parmer 4900592
The DataChart 1250 is a powerful, compact digital recorder with intuitive operation, flexible input configuration and large memory capability. Combining small package size with extreme flexibility make the DataChart 1250 a powerful, yet economical, digital data recording system. FEATURES
  • 2 universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTD's as well as frequency and pulse inputs
  • 4 internal alarm set points
  • 2 alarm relay outputs and 1 digital control input
  • Maximum sample storage rate of 100 samples per second per channel
  • Compact Flash cards up to 2 Gigabyte size
  • Catalog No. 50-631-720