Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare 2D Protein Extraction Buffer-V

Designed to produce better spot resolution for 2D gel analysis. GE Healthcare 2D Protein Extraction Buffer-V offers a convenient way to prepare high-quality protein lysates. Suitable for use with 2D or 1D electrophoresis, liquid chromatography, protein expression and mass spectrometry applications.

Manufacturer:  GE Healthcare 28943527

Catalog No. 45-002-415

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Efficient cell lysis and protein extraction are key steps for achieving high-quality results in downstream applications. 2D Protein Extraction Buffer-V offers a convenient way to prepare high-quality protein lysates. This buffer is a modification of well-studied protein solubilization and IPG-strip rehydration buffers. It is designed to produce high spot resolution for 2D gel analysis.
  • Buffer is available as a dry powder, which is rehydrated with the provided diluent
  • Contains urea, thiourea and CHAPS with Diluent-II
  • Compatible with CyDye™ DIGE Fluors
  • Extraction Buffer-V is suitable for stronger solubilization effects
Specifications

Specifications

For 50mL sample solution
This study presents a comparison between different protein extraction buffers to investigate the extraction capabilities and differences between the buffers
Rat liver proteins were extracted using 2-D Protein Extraction Buffer-I to -VI, as well as a standard 2-D lysis reference buffer
Contains Urea, Thiourea and CHAPS
30mL diluent supplied with the dry buffer component
Instructions supplied with the buffer
2-D Protein Extraction Buffers at 15°C to 30°C
DILUENTS at 2°C to 8°C
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit