Alfa Aesar™ (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one, 95%

Manufacturer: Alfa Aesar™ H3287506

Quantity 5g

 View more versions of this product

Catalog No. AAH3287506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14575-25-8
    Molecular Formula C21H21NO
    Formula Weight 303.41
    MDL Number MFCD06409497
    Synonym 4-(1,3,3-Trimethyl-2-indolinylidene)crotonophenone; (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-1,3-dihydro-2H-indol-2-ylidene)but-2-en-1-one
    Chemical Name or Material (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one

    Specifications

    Formula Weight 303.41
    Molecular Formula C21H21NO
    MDL Number MFCD06409497
    CAS 14575-25-8
    Synonym 4-(1,3,3-Trimethyl-2-indolinylidene)crotonophenone; (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-1,3-dihydro-2H-indol-2-ylidene)but-2-en-1-one
    Melting Point 156° to 162°C
    Quantity 5g
    Chemical Name or Material (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit