Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2E,5E)-2,5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
261360-66-1
C27H34N2O
402.58
MFCD06658953
KKLBPVXKMBLCQX-BLVCXSLXSA-N
cyclopentanone, 2,5-bis 4-diethylamino phenyl methylene,2e,5e-2,5-bis 4-diethylamino phenyl methylene cyclopentanone,2,5-bis 4-diethylamino benzylidene cyclopentanone,2,5-bis 4-diethylaminobenzylidene cyclopentanone 17,2,5-bis e-4-diethylamino benzylidene cyclopentanone,2e 5e-2,5-bis 4-diethylamino benzylidene cyclopentanone,2e,5e-2,5-bis 4-diethylamino benzylidene cyclopentanone,2,5-di e-1-4-diethylaminophenyl methylidene-1-cyclopentanone,2e,5e-2,5-bis 4-diethylamino phenyl methylidene cyclopentan-1-one
5702661
(2E,5E)-2,5-bis[[4-(diethylamino)phenyl]methylidene]cyclopentan-1-one
CCN(CC)C1=CC=C(C=C1)C=C2CCC(=CC3=CC=C(C=C3)N(CC)CC)C2=O
261360-66-1
C27H34N2O
402.58
MFCD06658953
KKLBPVXKMBLCQX-BLVCXSLXSA-N
cyclopentanone, 2,5-bis 4-diethylamino phenyl methylene,2e,5e-2,5-bis 4-diethylamino phenyl methylene cyclopentanone,2,5-bis 4-diethylamino benzylidene cyclopentanone,2,5-bis 4-diethylaminobenzylidene cyclopentanone 17,2,5-bis e-4-diethylamino benzylidene cyclopentanone,2e 5e-2,5-bis 4-diethylamino benzylidene cyclopentanone,2e,5e-2,5-bis 4-diethylamino benzylidene cyclopentanone,2,5-di e-1-4-diethylaminophenyl methylidene-1-cyclopentanone,2e,5e-2,5-bis 4-diethylamino phenyl methylidene cyclopentan-1-one
5702661
(2E,5E)-2,5-bis[[4-(diethylamino)phenyl]methylidene]cyclopentan-1-one
CCN(CC)C1=CC=C(C=C1)C=C2CCC(=CC3=CC=C(C=C3)N(CC)CC)C2=O
Melting Point 186°C to 189°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Red
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 402.58
Physical Form Powder
Percent Purity 95%
Chemical Name or Material (2E, 5E)-2, 5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning