Learn More
2',7'-Dichlorofluorescein, Thermo Scientific Chemicals
Description
2',7'-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2′,7′-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.
Solubility
Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.
Notes
As a fluorescent indicatorIncompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C (decomposition) |
Quantity | 5 g |
Solubility Information | Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water. |
Formula Weight | 401.2 |
Chemical Name or Material | 2′, 7′-Dichlorofluorescein |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H319 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 200-968-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.