Alfa Aesar™ (2H)1,4-Benzothiazin-3(4H)-one, 99%

Manufacturer: Alfa Aesar™ A1109122

Quantity 100g

 View more versions of this product

Catalog No. AAA1109122

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 5325-20-2
    Molecular Formula C8H7NOS
    Formula Weight 165.22
    MDL Number MFCD00006698
    Beilstein 131642
    Chemical Name or Material (2H)1,4-Benzothiazin-3 (4H)-one

    Specifications

    Formula Weight 165.22
    Molecular Formula C8H7NOS
    MDL Number MFCD00006698
    CAS 5325-20-2
    Beilstein 131642
    Melting Point 176° to 178°C
    Quantity 100g
    Chemical Name or Material (2H)1,4-Benzothiazin-3 (4H)-one
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit