Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (2H)1,4-Benzothiazin-3(4H)-one, 99%

Manufacturer:  Alfa Aesar™ A1109114

Quantity 25g

 View more versions of this product

Catalog No. AAA1109114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

5325-20-2
165.22
131642
C8H7NOS
MFCD00006698
(2H)1,4-Benzothiazin-3 (4H)-one

Specifications

176° to 178°C
5325-20-2
165.22
131642
99%
25g
C8H7NOS
MFCD00006698
(2H)1,4-Benzothiazin-3 (4H)-one
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit