Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene, 95%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
71042-54-1
C31H28P2
462.513
MFCD00085365
CDJHPMXMJUCLPA-WDHYYQACSA-N
2s,3s-+-2,3-bis diphenylphosphino bicyclo 2.2.1 hept-5-ene,2s,3s-norphos,2s,3s-+-norphos,2s,3s-3-diphenylphosphanyl bicyclo 2.2.1 hept-5-en-2-yl diphenylphosphane,phosphine,1,1'-1r,2s,3s,4s-bicyclo 2.2.1 hept-5-ene-2,3-diylbis 1,1-diphenyl
133126730
[(1R,2S,3S)-2-diphenylphosphanyl-3-bicyclo[2.2.1]hept-5-enyl]-diphenylphosphane
C1C2C=CC1C(C2P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6
71042-54-1
C31H28P2
462.513
MFCD00085365
CDJHPMXMJUCLPA-WDHYYQACSA-N
2s,3s-+-2,3-bis diphenylphosphino bicyclo 2.2.1 hept-5-ene,2s,3s-norphos,2s,3s-+-norphos,2s,3s-3-diphenylphosphanyl bicyclo 2.2.1 hept-5-en-2-yl diphenylphosphane,phosphine,1,1'-1r,2s,3s,4s-bicyclo 2.2.1 hept-5-ene-2,3-diylbis 1,1-diphenyl
133126730
[(1R,2S,3S)-2-diphenylphosphanyl-3-bicyclo[2.2.1]hept-5-enyl]-diphenylphosphane
C1C2C=CC1C(C2P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6
Packaging Glass bottle
Color White
Quantity 100mg
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 462.52
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (2R, 3R)-(-)-2, 3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene
Show More Show Less