Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3S)-1-Carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene, 95%, Acros Organics

Click to view available options
Quantity:
1g
193338-30-6
C8H10O4
170.164
MFCD12910462
YZZIXSUTJHCCFT-NKWVEPMBSA-N
5s,6r-5,6-dihydroxy-4-methylcyclohexa-1,3-diene-1-carboxylic acid,5s,6r-5,6-dihydroxy-4-methylcyclohexa-1,3-dienecarboxylic acid,2r,3s-1-carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene,2r,3s-1-carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene, 95,1,3-cyclohexadiene-1-carboxylicacid, 5,6-dihydroxy-4-methyl-, 5s,6r
2734009
(5S,6R)-5,6-dihydroxy-4-methylcyclohexa-1,3-diene-1-carboxylic acid
CC1=CC=C(C(C1O)O)C(=O)O
193338-30-6
C8H10O4
170.164
MFCD12910462
YZZIXSUTJHCCFT-NKWVEPMBSA-N
5s,6r-5,6-dihydroxy-4-methylcyclohexa-1,3-diene-1-carboxylic acid,5s,6r-5,6-dihydroxy-4-methylcyclohexa-1,3-dienecarboxylic acid,2r,3s-1-carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene,2r,3s-1-carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene, 95,1,3-cyclohexadiene-1-carboxylicacid, 5,6-dihydroxy-4-methyl-, 5s,6r
2734009
(5S,6R)-5,6-dihydroxy-4-methylcyclohexa-1,3-diene-1-carboxylic acid
CC1=CC=C(C(C1O)O)C(=O)O
Quantity 1g
Color White to Brown
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 170.16
Physical Form Fine Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (2R,3S)-1-Carboxy-2,3-dihydroxy-4-methylcyclohexa-4,6-diene, 95%