Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Acros Organics
Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
Specifications
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 500mg |
Color | Amber to Colorless |
Enantiomeric Excess | 97.5% min. (GC) |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Formula Weight | 259.3 |
Physical Form | Viscous Liquid or Waxy Mass |
Show More |
Safety and Handling
TSCA : TSCA