Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,5R)-2,5-Hexanediol, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
17299-07-9
C6H14O2
118.18
OHMBHFSEKCCCBW-PHDIDXHHSA-N
2r,5r-2,5-hexanediol,2r,5r-hexane-2,5-diol,2,5-hexanediol, 2r,5r,2r,5r---2,5-hexanediol,hx2,2r,5r-hexanediol,2r,5r-2,5-dihydroxyhexane,2r,5r-2,5-hexanediol sum of enantiomers, gc
2733360
(2R,5R)-hexane-2,5-diol
CC(CCC(C)O)O
17299-07-9
C6H14O2
118.18
OHMBHFSEKCCCBW-PHDIDXHHSA-N
2r,5r-2,5-hexanediol,2r,5r-hexane-2,5-diol,2,5-hexanediol, 2r,5r,2r,5r---2,5-hexanediol,hx2,2r,5r-hexanediol,2r,5r-2,5-dihydroxyhexane,2r,5r-2,5-hexanediol sum of enantiomers, gc
2733360
(2R,5R)-hexane-2,5-diol
CC(CCC(C)O)O
Melting Point 50°C to 53°C
Boiling Point 212°C to 215°C
Flash Point 101°C
Specific Rotation -34.5
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Formula Weight 118.18
Physical Form Crystalline Powder or Needles
Percent Purity 95%,98% ee
Chemical Name or Material (2R, 5R)-2, 5-Hexanediol, 98% ee
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning