Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
250mg
500mg
Packaging:
Glass bottle
441044-14-0
C18H28N2O5
352.431
QMLALKYUTWPCAI-WSZWBAFRSA-N
2s,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid benzylamine salt,2s,4s-1-tert-butoxycarbonyl-4-hydroxypiperidine-2-carboxylic acid; benzylamine,phenylmethanamine 2s,4s-1-tert-butoxycarbonyl-4-hydroxypiperidine-2-carboxylate,2s,4s-1-tert-butyl oxycarbonyl-4-hydroxypiperidine-2-carboxylic acid, phe nylmethylamine,2s,4s-4-hydroxy-1-2-methylpropan-2-yl oxycarbonyl piperidine-2-carboxylic acid; phenylmethanamine
2734505
(2S,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid;phenylmethanamine
CC(C)(C)OC(=O)N1CCC(CC1C(=O)O)O.C1=CC=C(C=C1)CN
441044-14-0
C18H28N2O5
352.431
QMLALKYUTWPCAI-WSZWBAFRSA-N
2s,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid benzylamine salt,2s,4s-1-tert-butoxycarbonyl-4-hydroxypiperidine-2-carboxylic acid; benzylamine,phenylmethanamine 2s,4s-1-tert-butoxycarbonyl-4-hydroxypiperidine-2-carboxylate,2s,4s-1-tert-butyl oxycarbonyl-4-hydroxypiperidine-2-carboxylic acid, phe nylmethylamine,2s,4s-4-hydroxy-1-2-methylpropan-2-yl oxycarbonyl piperidine-2-carboxylic acid; phenylmethanamine
2734505
(2S,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid;phenylmethanamine
CC(C)(C)OC(=O)N1CCC(CC1C(=O)O)O.C1=CC=C(C=C1)CN
Enantiomeric Excess 97.5% (HPLC) min.
Packaging Glass bottle
Quantity 500mg
Infrared Spectrum Authentic
Color White
Assay Percent Range 95%
Formula Weight 352.43
Physical Form Crystalline Powder
Percent Purity ≥94% (HPLC)
Chemical Name or Material (2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt