Alfa Aesar™ (2S,4S)-(+)-Pentanediol, 99%, ee 99%

Manufacturer: Alfa Aesar™ H3601318

Quantity 50g

 View more versions of this product

Catalog No. AAH3601318

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 72345-23-4
    Formula Weight 104.15
    MDL Number MFCD00064276
    Beilstein 4123490
    Synonym (2S,4S)-(+)-2,4-Dihydroxypentane
    Chemical Name or Material (2S,4S)-(+)-Pentanediol, ee 99%

    Specifications

    Formula Weight 104.15
    Melting Point 45° to 48°C
    CAS 72345-23-4
    Color White
    Sensitivity Hygroscopic
    Physical Form Crystalline Powder
    MDL Number MFCD00064276
    Beilstein 4123490
    Synonym (2S,4S)-(+)-2,4-Dihydroxypentane
    Quantity 50g
    Chemical Name or Material (2S,4S)-(+)-Pentanediol, ee 99%
    Assay 99%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit