Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
1314246-02-0
C17H25O2P
292.35
UKBFKBMQNUOKTD-KBPBESRZSA-N
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,s,s-et-rajphos,2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane,2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane,trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
16218309
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
1314246-02-0
C17H25O2P
292.35
UKBFKBMQNUOKTD-KBPBESRZSA-N
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,s,s-et-rajphos,2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane,2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane,trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane,2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
16218309
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
Flash Point >110°C
Quantity 100mg
NMR 96.0% min.
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Formula Weight 292.35
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane
Show More Show Less