Alfa Aesar™ (2S)-(+)-Glycidyl p-toluenesulfonate, 99%

Manufacturer: Alfa Aesar™ H5600006

Quantity 5g

 View more versions of this product

Catalog No. AAH5600006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70987-78-9
    Molecular Formula C10H12O4S
    Formula Weight 228.26
    MDL Number MFCD00064489
    UN Number UN3077
    Beilstein 3592143
    Synonym (2S)-(+)-Glycidyl tosylate
    Chemical Name or Material (2S)-(+)-Glycidyl p-toluenesulfonate

    Specifications

    Formula Weight 228.26
    UN Number UN3077
    Synonym (2S)-(+)-Glycidyl tosylate
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Flash Point 113°C (235°F)
    RTECS Number RR0510050
    Molecular Formula C10H12O4S
    MDL Number MFCD00064489
    CAS 70987-78-9
    Beilstein 3592143
    Health Hazard 2 GHS H Statement
    H350-H341-H318-H317
    May cause cancer.
    Suspected of causing genetic defects.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Melting Point 46° to 49°C
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material (2S)-(+)-Glycidyl p-toluenesulfonate
    Assay 99%

    GHS H Statement
    H350-H341-H318-H317
    May cause cancer.
    Suspected of causing genetic defects.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit