Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(1-Homopiperazinylmethyl)indole, 95%

Click to view available options
Quantity:
250mg
1g
5g
1183967-11-4
3-(1,4-Diazepan-1-ylmethyl)indole
1183967-11-4
3-(1,4-Diazepan-1-ylmethyl)indole
Quantity 250mg
UN Number UN3259
Beilstein 11950916
Formula Weight 229.32
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-(1-Homopiperazinylmethyl)indole

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures