Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(1-Naphthyl)acrylic acid, 98%

Click to view available options
Quantity:
10g
50g
250g
13026-12-5
C13H10O2
198.221
MFCD00014317
WPXMLUUYWNHQOR-HJWRWDBZSA-N
naphthalene-1-acrylic acid,z-3-1-naphthyl acrylic acid,z-3-naphthalen-1-yl-acrylic acid,z-3-naphthalen-1-yl acrylic acid,2z-3-naphthalen-1-yl prop-2-enoic acid
12158295
(Z)-3-naphthalen-1-ylprop-2-enoic acid
C1=CC=C2C(=C1)C=CC=C2C=CC(=O)O
13026-12-5
C13H10O2
198.221
MFCD00014317
WPXMLUUYWNHQOR-HJWRWDBZSA-N
naphthalene-1-acrylic acid,z-3-1-naphthyl acrylic acid,z-3-naphthalen-1-yl-acrylic acid,z-3-naphthalen-1-yl acrylic acid,2z-3-naphthalen-1-yl prop-2-enoic acid
12158295
(Z)-3-naphthalen-1-ylprop-2-enoic acid
C1=CC=C2C(=C1)C=CC=C2C=CC(=O)O
Applications
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Melting Point 210°C to 212°C
Quantity 10g
Beilstein 1868617
Solubility Information Slightly soluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3-(1-Naphthyl)acrylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-887-5

TSCA : No

Recommended Storage : Ambient temperatures