Alfa Aesar™ 3-(1-Piperazinyl)-1,2-benzisoxazole, 96%

Manufacturer: Alfa Aesar™ H50746MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50746MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87691-89-2
    Molecular Formula C11H13N3O
    Formula Weight 203.25
    MDL Number MFCD08060968
    Synonym 3-(1-Piperazino)-1,2-benzisoxazole
    Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisoxazole

    Specifications

    Formula Weight 203.25
    Molecular Formula C11H13N3O
    MDL Number MFCD08060968
    Synonym 3-(1-Piperazino)-1,2-benzisoxazole
    Melting Point 74° to 76°C
    Quantity 250mg
    CAS 87691-89-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisoxazole
    Assay 96%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit