Learn More
Sigma Organic Chemistry 3- 1-Pyrrolidinyl propa | 50mg | MFCD06801279
Supplier: Sigma Organic Chemistry CDS00232550MG

3- 1-Pyrrolidinyl propa , 50mg
About this Item:
MDL #: MFCD06801279
Molecular Weight: 179.64
UNSPSC Code: 12352200
Chemical Formula: C7H14ClNO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.