Alfa Aesar™ 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H2880103

Quantity 1g

 View more versions of this product

Catalog No. AAH2880103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1073353-61-3
    Molecular Formula C17H24BNO3
    Formula Weight 301.19
    MDL Number MFCD09027086
    Synonym 3-(Pyrrolidine-1-carbonyl)phenylboronic acid pinacol ester; 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]pyrrolidine
    Chemical Name or Material 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 301.19
    Molecular Formula C17H24BNO3
    MDL Number MFCD09027086
    CAS 1073353-61-3
    Synonym 3-(Pyrrolidine-1-carbonyl)phenylboronic acid pinacol ester; 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]pyrrolidine
    Melting Point 96° to 98°C
    Quantity 1g
    Chemical Name or Material 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit