Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(1H-Tetrazol-5-yl)aniline, 96%

Manufacturer:  Alfa Aesar™ B2250209

Quantity 10g

 View more versions of this product

Catalog No. AAB2250209


  Check Availability
Add to cart

Specifications

Description

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

73732-51-1
161.17
5-(3-Aminophenyl)tetrazole
C7H7N5
MFCD01219561
3-(1H-Tetrazol-5-yl)aniline

Specifications

202° to 205°C
73732-51-1
161.17
5-(3-Aminophenyl)tetrazole
96%
10g
C7H7N5
MFCD01219561
3-(1H-Tetrazol-5-yl)aniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BY9276000

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'