Alfa Aesar™ 3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

Manufacturer: Alfa Aesar™ 4653922

Quantity 100g

 View more versions of this product

Catalog No. AA4653922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 903876-45-9
    Molecular Formula C19H30O2
    Formula Weight 290.45
    MDL Number MFCD22380703
    Synonym IBCHA; Acrylic acid 3-isobornylcyclohexyl ester
    Chemical Name or Material 3- (2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

    Specifications

    Formula Weight 290.45
    Health Hazard 2 GHS H Statement
    H334-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Color Pale brown
    TSCA TSCA
    Molecular Formula C19H30O2
    MDL Number MFCD22380703
    CAS 903876-45-9
    Synonym IBCHA; Acrylic acid 3-isobornylcyclohexyl ester
    Health Hazard 1 Danger
    Recommended Storage 2°C to 8°C
    Flash Point >149°C (300°F)
    Quantity 100g
    Chemical Name or Material 3- (2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

    GHS H Statement
    H334-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    2°C to 8°C

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit