Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3(2),6,6-Trimethylfulvene, mixture of isomers, tech. 90%

Click to view available options
Quantity:
1g
5g
1217863-24-5
C6H12
MFCD02683073
1217863-24-5
C6H12
MFCD02683073
Quantity 1g
UN Number UN3295
Formula Weight 120.19
Percent Purity 90%
Grade Technical
Chemical Name or Material 3(2),6,6-Trimethylfulvene, mixture of isomers

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.