Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3(2),6,6-Trimethylfulvene, mixture of isomers, tech. 90%
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 5g |
UN Number | UN3295 |
Formula Weight | 120.19 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 3(2),6,6-Trimethylfulvene, mixture of isomers |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures