Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog Number AAB2315922
Click to view available options
Quantity:
25 g
100 g
500 g
3069-29-2
C8H22N2O2Si
206.36
MFCD00054776
MQWFLKHKWJMCEN-UHFFFAOYSA-N
n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
76482
N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine
CO[Si](C)(CCCNCCN)OC
3069-29-2
C8H22N2O2Si
206.36
MFCD00054776
MQWFLKHKWJMCEN-UHFFFAOYSA-N
n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
76482
N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine
CO[Si](C)(CCCNCCN)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 265°C
Quantity 100 g
UN Number UN2735
Beilstein 2074259
Flash Point 90°C (194°F)
Formula Weight 206.36
Percent Purity 97%
Odor Amine-like
Refractive Index 1.449
Sensitivity Air and moisture sensitive
Density 0.968
Chemical Name or Material 3-(2-Aminoethylamino)propylmethyldimethoxysilane
Show More Show Less
Hazard Category H227-H302-H315-H317-H318
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 221-336-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit