Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[2-(Benzoyloxy)phenyl]propionic acid, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
59725-59-6
C16H14O4
270.284
MFCD00052018
WZLUNIRPOQOEJR-UHFFFAOYSA-N
3-2-benzoyloxy phenyl propanoic acid,3-2-benzoyloxy phenyl propionic acid,maybridge1_007224,acmc-20anb0,3-2-benzoyloxyphenyl propanoic acid,benzenepropanoic acid,2-benzoyloxy,3-2-phenylcarbonyloxyphenyl propanoic acid
2732579
3-(2-benzoyloxyphenyl)propanoic acid
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2CCC(=O)O
59725-59-6
C16H14O4
270.284
MFCD00052018
WZLUNIRPOQOEJR-UHFFFAOYSA-N
3-2-benzoyloxy phenyl propanoic acid,3-2-benzoyloxy phenyl propionic acid,maybridge1_007224,acmc-20anb0,3-2-benzoyloxyphenyl propanoic acid,benzenepropanoic acid,2-benzoyloxy,3-2-phenylcarbonyloxyphenyl propanoic acid
2732579
3-(2-benzoyloxyphenyl)propanoic acid
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2CCC(=O)O
Quantity 1g
Formula Weight 270.29
Percent Purity 97%
Chemical Name or Material 3-[2-(Benzoyloxy)phenyl]propionic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning