Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Furyl)-2-phenylpropenal, 97%, Thermo Scientific Chemicals

Catalog No. AAL0622014
Click to view available options
Quantity:
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57568-60-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00036566
InChI Key JPESOGFYFXAURP-XFXZXTDPSA-N
Synonym 3-2-furyl-2-phenylpropenal, 3-2'-furyl-2-phenyl-2-propenal, 3-2-furyl-2-phenyl propenal, z-3-furan-2-yl-2-phenylprop-2-enal, 2z-3-furan-2-yl-2-phenylprop-2-enal
PubChem CID 6537126
IUPAC Name (Z)-3-(furan-2-yl)-2-phenylprop-2-enal
SMILES O=C\C(=C\C1=CC=CO1)C1=CC=CC=C1

Specifications

Melting Point 57°C to 60°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 5923
Sensitivity Air Sensitive
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)-2-phenylpropenal
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.