Alfa Aesar™ 3-(2-Methylphenoxy)-1,2-propanediol, 98%

Manufacturer: Alfa Aesar™ B2192822

Quantity 100g

 View more versions of this product

Catalog No. AAB2192822

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 59-47-2
    Molecular Formula C10H14O3
    Formula Weight 182.22
    MDL Number MFCD00004718
    Merck Index 14,5850
    Synonym Mephenesin; 3-(o-Tolyloxy)-1,2-propanediol
    Chemical Name or Material 3-(2-Methylphenoxy)-1,2-propanediol

    Specifications

    Formula Weight 182.22
    Solubility Information Soluble in alcohol
    Melting Point 69° to 71°C
    Quantity 100g
    Molecular Formula C10H14O3
    MDL Number MFCD00004718
    CAS 59-47-2
    Merck Index 14,5850
    Synonym Mephenesin; 3-(o-Tolyloxy)-1,2-propanediol
    Health Hazard 1 Danger
    Boiling Point 153° to 154°C (4mmHg)
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-(2-Methylphenoxy)-1,2-propanediol
    Assay 98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit