Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(2-Naphthyl)acrylic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
51557-26-7
C13H10O2
198.221
MFCD00209666
KWGPBDBAAXYWOJ-VURMDHGXSA-N
naphthalene-2-acrylic acid,z-3-2-naphthyl prop-2-enoic acid,z-3-naphthalen-2-ylprop-2-enoic acid
1712266
(Z)-3-naphthalen-2-ylprop-2-enoic acid
C1=CC=C2C=C(C=CC2=C1)C=CC(=O)O
51557-26-7
C13H10O2
198.221
MFCD00209666
KWGPBDBAAXYWOJ-VURMDHGXSA-N
naphthalene-2-acrylic acid,z-3-2-naphthyl prop-2-enoic acid,z-3-naphthalen-2-ylprop-2-enoic acid
1712266
(Z)-3-naphthalen-2-ylprop-2-enoic acid
C1=CC=C2C=C(C=CC2=C1)C=CC(=O)O
Quantity 25g
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3-(2-Naphthyl)acrylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures