Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(2-Naphthyl)-DL-alanine, 98%

Click to view available options
Quantity:
250mg
1g
5g
14108-60-2
C13H13NO2
215.252
MFCD00070216
JPZXHKDZASGCLU-UHFFFAOYSA-N
2-amino-3-naphthalen-2-yl propanoic acid,3-2-naphthyl-dl-alanine,h-dl-2-nal-oh,2-naphthalenealanine,2-amino-3-naphthalen-2-yl-propionic acid,dl-3-2-naphthyl-alanine,3-2-naphthyl alanine,beta-naphtyl alanine,acmc-209m4i,acmc-209p7c
250399
2-amino-3-naphthalen-2-ylpropanoic acid
C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N
14108-60-2
C13H13NO2
215.252
MFCD00070216
JPZXHKDZASGCLU-UHFFFAOYSA-N
2-amino-3-naphthalen-2-yl propanoic acid,3-2-naphthyl-dl-alanine,h-dl-2-nal-oh,2-naphthalenealanine,2-amino-3-naphthalen-2-yl-propionic acid,dl-3-2-naphthyl-alanine,3-2-naphthyl alanine,beta-naphtyl alanine,acmc-209m4i,acmc-209p7c
250399
2-amino-3-naphthalen-2-ylpropanoic acid
C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N
Formula Weight 215.25
Physical Form Powder
Percent Purity 98%
Color White
Quantity 1g
Chemical Name or Material 3-(2-Naphthyl)-DL-alanine

storageNote1 : 2° to 8°C