Alfa Aesar™ 3-(2-Pyrrolidinyl)piperidine, 94%

Manufacturer: Alfa Aesar™ H26049MC

Quantity 100mg

 View more versions of this product

Catalog No. AAH26049MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 858262-19-8
    Molecular Formula C9H18N2
    Formula Weight 154.26
    MDL Number MFCD08277606
    UN Number UN3259
    Synonym 2-(3-Piperidinyl)pyrrolidine; 3-(2-Pyrrolidino)piperidine
    Chemical Name or Material 3-(2-Pyrrolidinyl)piperidine

    Specifications

    Formula Weight 154.26
    UN Number UN3259
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 100mg
    Molecular Formula C9H18N2
    MDL Number MFCD08277606
    Synonym 2-(3-Piperidinyl)pyrrolidine; 3-(2-Pyrrolidino)piperidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    CAS 858262-19-8
    Chemical Name or Material 3-(2-Pyrrolidinyl)piperidine
    Assay 94%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit