Alfa Aesar™ 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H3032314

Quantity 25g

 View more versions of this product

Catalog No. AAH3032314

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C18H31BO3Si
    Formula Weight 334.34
    MDL Number MFCD12546185
    Synonym 4,4,5,5-Tetramethyl-2-[3-(2-trimethylsiloxyphenyl)propyl]-1,3,2-dioxaborolane
    Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester

    Specifications

    Formula Weight 334.34
    Synonym 4,4,5,5-Tetramethyl-2-[3-(2-trimethylsiloxyphenyl)propyl]-1,3,2-dioxaborolane
    Quantity 25g
    Molecular Formula C18H31BO3Si
    MDL Number MFCD12546185
    Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit