Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)indocarbocyanine Sodium Salt 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
120724-84-7
C31H39N2NaO6S2
622.771
MFCD16621066
SBQIZSUJAUKSIJ-UHFFFAOYSA-M
2-[3-[3,3-Dimethyl-1-(4-sulfobutyl)-1,3-dihydroindol-2-ylidene]propenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Hydroxide Inner Salt Sodium Salt, 3-[3,3-Dimethyl-1-(4-sulfonatobutyl)indolin-2-ylidene]-1-[3,3-dimethyl-1-(4-sulfonatobutyl)indolium-2-yl]-
133556299
sodium;4-[(2E)-2-[(E)-3-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium-2-yl]prop-2-enylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate
CC1(C2=CC=CC=C2[N+](=C1C=CC=C3C(C4=CC=CC=C4N3CCCCS(=O)(=O)[O-])(C)C)CCCCS(=O)(=O)[O-])C.[Na+]
120724-84-7
C31H39N2NaO6S2
622.771
MFCD16621066
SBQIZSUJAUKSIJ-UHFFFAOYSA-M
2-[3-[3,3-Dimethyl-1-(4-sulfobutyl)-1,3-dihydroindol-2-ylidene]propenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Hydroxide Inner Salt Sodium Salt, 3-[3,3-Dimethyl-1-(4-sulfonatobutyl)indolin-2-ylidene]-1-[3,3-dimethyl-1-(4-sulfonatobutyl)indolium-2-yl]-
133556299
sodium;4-[(2E)-2-[(E)-3-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium-2-yl]prop-2-enylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate
CC1(C2=CC=CC=C2[N+](=C1C=CC=C3C(C4=CC=CC=C4N3CCCCS(=O)(=O)[O-])(C)C)CCCCS(=O)(=O)[O-])C.[Na+]
Quantity 1g
Color Undesignated
Formula Weight 622.77
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)indocarbocyanine Sodium Salt