Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3-Trifluoropropionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
100mL
Packaging:
Glass Bottle
2516-99-6
C3H3F3O2
128.05
MFCD00153292
KSNKQSPJFRQSEI-UHFFFAOYSA-N
3,3,3-trifluoropropionic acid,3,3,3-trifluoro-propionic acid,propanoic acid, 3,3,3-trifluoro,tfpa,acmc-209ghu,trifluoromethylacetic acid,trifluoromethyl acetic acid,ksc203o6p,3,3,3,-trifluoropropionic acid,3,3,3-tri-fluoropropionic acid
2777972
3,3,3-trifluoropropanoic acid
C(C(=O)O)C(F)(F)F
2516-99-6
C3H3F3O2
128.05
MFCD00153292
KSNKQSPJFRQSEI-UHFFFAOYSA-N
3,3,3-trifluoropropionic acid,3,3,3-trifluoro-propionic acid,propanoic acid, 3,3,3-trifluoro,tfpa,acmc-209ghu,trifluoromethylacetic acid,trifluoromethyl acetic acid,ksc203o6p,3,3,3,-trifluoropropionic acid,3,3,3-tri-fluoropropionic acid
2777972
3,3,3-trifluoropropanoic acid
C(C(=O)O)C(F)(F)F
Packaging Glass Bottle
Refractive Index 1.3320 to 1.334
Flash Point 92°C
Boiling Point 146°C
Specific Gravity 1.45
Melting Point 9.7°C
Quantity 1mL
Infrared Spectrum Authentic
Density 1.4500g/mL
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula F3CCH2COOH
Formula Weight 128.05
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3, 3, 3-Trifluoropropionic acid
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger