Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3,3-Triphenylpropionic acid, 98%

Click to view available options
Quantity:
5g
25g
900-91-4
C21H18O2
302.373
MFCD00002713
XMSJLUKCGWQAHO-UHFFFAOYSA-N
3,3,3-triphenylpropionic acid,tritylacetic acid,3,3,3-triphenyl propanoic acid,2-tritylacetic acid,pubchem12502,acmc-209r4v,2-triphenylmethylacetic acid,2-triphenylmethyl acetic acid,3,3 3-triphenyl propionic acid
136696
3,3,3-triphenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)(C2=CC=CC=C2)C3=CC=CC=C3
900-91-4
C21H18O2
302.373
MFCD00002713
XMSJLUKCGWQAHO-UHFFFAOYSA-N
3,3,3-triphenylpropionic acid,tritylacetic acid,3,3,3-triphenyl propanoic acid,2-tritylacetic acid,pubchem12502,acmc-209r4v,2-triphenylmethylacetic acid,2-triphenylmethyl acetic acid,3,3 3-triphenyl propionic acid
136696
3,3,3-triphenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)(C2=CC=CC=C2)C3=CC=CC=C3
Applications
3,3,3-Triphenylpropionic acid is used as pharmaceutical intermediates.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Quantity 25g
Melting Point 180°C to 183°C
Beilstein 2057448
Formula Weight 302.38
Percent Purity 98%
Chemical Name or Material 3,3,3-Triphenylpropionic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures