Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-(3,4-Dimethoxyphenyl)propionic acid, 99%, Thermo Scientific™
Click to view available options
50g; Glass bottle
Specifications
CAS | 2107-70-2 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C11H14O4 |
Linear Formula | (CH3O)2C6H3CH2CH2CO2H |
Quantity | 50g |
MDL Number | MFCD00002774 |
Show More |
Safety and Handling
EINECSNumber | 218-288-3 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction