Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Acros Organics

Click to view available options
Quantity:
2g
10g
2455-14-3
C28H40O2
408.63
GQIGHOCYKUBBOE-UHFFFAOYSA-N
3,3',5,5'-tetra-tert-butyldiphenoquinone,3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone,3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione,unii-4m92z38q9a,3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone,bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl,2,2',6,6'-tetra-tert-butyldiphenylquinone,2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one,pubchem13798,rarechem bw gc 0005
225283
2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C
2455-14-3
C28H40O2
408.63
GQIGHOCYKUBBOE-UHFFFAOYSA-N
3,3',5,5'-tetra-tert-butyldiphenoquinone,3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone,3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione,unii-4m92z38q9a,3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone,bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl,2,2',6,6'-tetra-tert-butyldiphenylquinone,2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one,pubchem13798,rarechem bw gc 0005
225283
2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C
Melting Point 247.0°C to 248.0°C
Quantity 10g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 408.63
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetra-tert-butyldiphenoquinone

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 219-527-4