Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3',5,5'-Tetrabromobisphenol A, 97%

Click to view available options
Quantity:
100g
2500g
79-94-7
C15H12Br4O2
543.875
MFCD00013962
VEORPZCZECFIRK-UHFFFAOYSA-N
tetrabromobisphenol a,3,3',5,5'-tetrabromobisphenol a,bromdian,4,4'-propane-2,2-diyl bis 2,6-dibromophenol,4,4'-isopropylidenebis 2,6-dibromophenol,firemaster bp 4a,tetrabromodian,2,2-bis 3,5-dibromo-4-hydroxyphenyl propane,fire guard 2000,great lakes ba-59p
6618
CHEBI:33217
2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC(=C(C(=C1)Br)O)Br)C2=CC(=C(C(=C2)Br)O)Br
79-94-7
C15H12Br4O2
543.875
MFCD00013962
VEORPZCZECFIRK-UHFFFAOYSA-N
tetrabromobisphenol a,3,3',5,5'-tetrabromobisphenol a,bromdian,4,4'-propane-2,2-diyl bis 2,6-dibromophenol,4,4'-isopropylidenebis 2,6-dibromophenol,firemaster bp 4a,tetrabromodian,2,2-bis 3,5-dibromo-4-hydroxyphenyl propane,fire guard 2000,great lakes ba-59p
6618
CHEBI:33217
2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC(=C(C(=C1)Br)O)Br)C2=CC(=C(C(=C2)Br)O)Br
Density 1.76
Quantity 100g
Melting Point 179°C to 182°C
Odor Odorless
UN Number UN3077
Formula Weight 543.87
Percent Purity 97%
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 201-236-9

RTECSNumber : SM0894500

TSCA : Yes

Recommended Storage : Ambient temperatures