Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3′,5,5′-Tetrachlorodiphenyl Disulfide 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
137897-99-5
C12H6Cl4S2
356.1
MFCD01862261
JMQANWHMOHXBEA-UHFFFAOYSA-N
Bis(3,5-dichlorophenyl) Disulfide
2735516
1,3-dichloro-5-[(3,5-dichlorophenyl)disulfanyl]benzene
C1=C(C=C(C=C1Cl)Cl)SSC2=CC(=CC(=C2)Cl)Cl
137897-99-5
C12H6Cl4S2
356.1
MFCD01862261
JMQANWHMOHXBEA-UHFFFAOYSA-N
Bis(3,5-dichlorophenyl) Disulfide
2735516
1,3-dichloro-5-[(3,5-dichlorophenyl)disulfanyl]benzene
C1=C(C=C(C=C1Cl)Cl)SSC2=CC(=CC(=C2)Cl)Cl
Color Yellow
Melting Point 67°C
Quantity 5g
Formula Weight 356.10
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material 3,3′,5,5′-Tetrachlorodiphenyl Disulfide

EINECSNumber : (3)-4544

TSCA : No